Skip to Content
Merck
All Photos(1)

Key Documents

XX3002502

Millipore

Filter Holder Gasket, PTFE

Accessories for filter holders for sample preparation.

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€127.00

€127.00


Estimated to ship on17 February 2025Details



Select a Size

Change View
10 EA
€127.00

About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

€127.00


Estimated to ship on17 February 2025Details


material

PTFE

Quality Level

diam.

25 mm

shipped in

ambient

Application

Accessories for filter holders for sample preparation.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service