Skip to Content
Merck
All Photos(1)

Key Documents

WMBSMT001

Milli-Q

Wall Mounting Bracket

For installation of RiOs 3, RiOs-DI® 3 and Direct-Q® 3 systems

Synonym(s):

Wall Mounting Bracket with Tap for RiOs, RiOs-DI® and Direct-Q® Systems

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€287.00

€287.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
€287.00

About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.13

€287.00


Please contact Customer Service for Availability

Request more information

feature

Saves lab bench space

packaging

pkg of 1 unit

compatibility

for use with Direct-Q® UV
for use with Direct-Q®
for use with RiOs-DI® UV
for use with RiOs-DI®
for use with RiOs-DI® Clinical
for use with RiOs

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Wall mounting bracket helps you to easily install the water purification system, anywhere you need pure water, on the benchtop or high on the wall, freeing your bench for crucial research work.

Application

Wall Mounting Bracket with Tap supports RiOs, RiOs-DI® and Direct-Q® Systems.

Features and Benefits

  • Increase access to water from your bench.
  • Saves more space.

Legal Information

DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany
RIOS-DI is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service