SPR0LSIA1
SmartPak® Purification Pack
Purifies pure and ultrapure water for Direct-Q® systems with low (<15 °C) feed water temperature
Synonym(s):
SmartPak® DQ3 Purification Pack LT (Low Temperature)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
feature
Produces ultrapure water directly from tap water
packaging
pkg of 1 unit
manufacturer/tradename
SmartPak®
resistivity
>18.0 MΩ-cm (at 25 degree celcius)
compatibility
for use with Direct-Q® UV
for use with Direct-Q®
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
The SmartPak® DQ3 Purification Pack LT contains a combination of selective purification media to produce Type 3 pure and Type 1 ultrapure water directly from tap water.
The SmartPak® DQ3 is an all-in-one twin cartridge removes trace impurities from tap water. The first cartridge combines pretreatment media and a reverse osmosis (RO) membrane. The second cartridge contains ion-exchange resins.
This pack contains exclusive Jetpore®, ultrapure grade mixed-bed ion-exchange resin to produce pure and ultrapure water directly from tap water.
The SmartPak® needs to be periodically replaced during maintenance of the system.
For use with Direct-Q® water purification systems with low (<15 °C) feed water temperature.
The SmartPak® DQ3 is an all-in-one twin cartridge removes trace impurities from tap water. The first cartridge combines pretreatment media and a reverse osmosis (RO) membrane. The second cartridge contains ion-exchange resins.
This pack contains exclusive Jetpore®, ultrapure grade mixed-bed ion-exchange resin to produce pure and ultrapure water directly from tap water.
The SmartPak® needs to be periodically replaced during maintenance of the system.
For use with Direct-Q® water purification systems with low (<15 °C) feed water temperature.
Application
This pack removes a variety of water contaminants (ions, organics, particles...).
Features and Benefits
- Pretreatment (activated carbon and depth filter).
- Reverse osmosis cartridge.
- Synthetic activated carbon.
- Virgin ion exchange resins.
- Care-free maintenance: easily installed and replaced.
- Certificate of Quality available online.
Legal Information
DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
SMARTPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service