Skip to Content
Merck
All Photos(1)

Documents

WBSH0500

Millipore

Immobilon Signal Enhancer for Immunodetection

Western Blotting, immunodetection

Synonym(s):

Western blot signal enhancer, signal enhancer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

Quality Level

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

detection method

chemiluminescent

shipped in

wet ice

General description

Immobilon® Signal Enhancer combines signal amplification and blocking in one ready-to-use reagent.
Signal Enhancing: Amplify low signal intensity Western blots without amplifying the noise
Save Primary Antibody: When the signal is strong, use Immobilon Signal Enhancer to reduce the amount of valuable primary antibody required

Application

Immobilon® Signal Enhancer is designed for use in Western blotting applications when low signal intensity is observed. It combines signal amplification and blocking in one ready-to-use reagent. Enhancement of the antibody-antigen reaction can produce a 2 to 10-fold increase in signal intensity over traditionally used reagents. The reagent is formulated for use in both membrane blocking and antibody dilution steps and can be used for traditional immunodetection or with the SNAP i.d. Protein Detection System. Immobilon Signal Enhancer is compatible with PVDF as well as nitrocellulose blotting membranes and can be used with chromogenic, chemiluminescent, and fluorescent detection methods.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service