Skip to Content
Merck
All Photos(3)

Key Documents

SYMBDTF04

Millipore

Steritest® Symbio Complete Drain Tray

For 3 canisters, FLEX pump, High Integration

Synonym(s):

Outlet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NB.13
Pricing and availability is not currently available.

material

316L stainless steel (Drain Tray Support)
EPDM rubber seal
polyphenylsulfone (PPSU) container

Agency

complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

General description

Steritest® Symbio complete drain tray includes stainless steel canister holder, plastic fluid collector, and seal. This tray is designed for 3 canisters and the usage of FLEX Pump in high integration. The versatile Steritest® Symbio FLEX Pump can be installed in either an isolator or a laminar flow hood.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service