Skip to Content
Merck
All Photos(2)

Key Documents

SYMBDSF01

Millipore

Steritest® Symbio drain tray support for 2 canisters, FLEX pump, low integration

This accessory streamlines the workflow and increases safety during sterility testing

Synonym(s):

Canister holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€798.00

€798.00


Estimated to ship on22 April 2025Details


Request more information

Select a Size

Change View
1 EACH
€798.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

€798.00


Estimated to ship on22 April 2025Details


Request more information

material

316L stainless steel support (Drain tray)
EPDM rubber seal

Agency

EP (2.6.1)
JP (4.06)
USP (71)

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio drain tray support includes the stainless steel canister holder and quick connector designed for 2 canisters FLEX pump in Low integration.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service