SNAP2FRMN01
SNAP id® 2.0 Midi Blot Holding Frame
Frame ensures membrane is horizontal allowing uniform Antibody distribution. The flat profile allows for compact space allocation for storage, shipping & waste management.
Select a Size
€149.00
Estimated to ship on06 May 2025Details
Select a Size
About This Item
€149.00
Estimated to ship on06 May 2025Details
Recommended Products
Product Name
SNAP id® 2.0 Mini Blot Holding Frame (single pack), The new Mini Blot Holders ensure the membrane is horizontal allowing uniform Antibody distribution. The flat profile alows for compact space allocation for storage, shipping & waste management.
Quality Level
manufacturer/tradename
SNAP id®
technique(s)
western blot: suitable
compatibility
for use with Commercially available blocking reagents
for use with Luminata Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01)
for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01)
for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001)
for use with commercially available detection reagents
detection method
chemiluminescent
colorimetric
fluorometric
shipped in
ambient
storage temp.
room temp
Related Categories
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service