Skip to Content
Merck
All Photos(1)

Key Documents

SLM-243-B

Sigma-Aldrich

DMEM/F-12

Complete medium, with ʟ-glutamine and 10% FBS, liquid, sterile-filtered, suitable for cell cutlure

Synonym(s):

DME/Nutrient Mixture F-12 Ham, DMEM Hams F12

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€215.00

€215.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 ML
€215.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71

€215.00


In StockDetails


Request a Bulk Order

Product Name

DMEM/F12 Complete with 2mM L-Glut and 10% FBS, 1:1 mixture of DMEM medium and Ham′s F12 medium, with 2mM L-Glut and 10% U.S. origin FBS

Quality Level

form

liquid

shipped in

ambient

Quality

pH= 7.2-7.5
Osmolality= 281-312
Endotoxin= ≤2 EU/mL
Mycoplasma= Negative
Sterility Tested= Negative for Growth
Appearance= Orange/ Red Clear Liquid
Cell Line Tested= ATDC5 Mouse Chondrogenic

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service