Skip to Content
Merck
All Photos(1)

Key Documents

M3942

SAFC

DMEM - high glucose

MegaCell, with sodium pyruvate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell cutlure

Synonym(s):

DMEM, DMEM Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€56.70

€56.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€56.70

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

€56.70


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

MegaCell Dulbecco′s Modified Eagle′s Medium, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
glucose: 4500 mg/L (Dextro)
sodium pyruvate: yes

shipped in

ambient

storage temp.

2-8°C

General description

MegaCell is a multifaceted media developed to reduce the amount of serum required for culturing mammalian cells in vitro. It reduces the supplementation of fetal bovine serum (FBS) or other serum from 10% to 3% and supports proliferative rates and maximal cell densities. MegaCell enables maintaining a long-term culture of non-fastidious cell lines.

Application

MegaCell Dulbecco′s Modified Eagle′s Medium has been used:
  • as a component of the M10 medium to culture CD133+ cells from biopsies of para-spinal and quadriceps muscle of Duchenne muscular dystrophy patients
  • as a component of the proliferation medium for the expansion and cryopreservation of human induced pluripotent stem (iPS) cells cultured without feeder cells and human embryonic (ES) cell-derived mesoangioblast-like cells
  • to culture the positive hybridomas during the production of mouse anti-chicken CD25 monoclonal antibodies

Reconstitution

Supplement with 4 mM L-glutamine (20 ml of 200 mM solution, G7513)

Legal Information

Sigma-Aldrich uses the MegaCell trademark pursuant to an agreement with Promega Corporation.
MegaCell is a trademark of Promega Corp.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shintaro Iwashita et al.
Bioscience reports, 35(4), doi:10-doi:10 (2015-07-17)
The BCNT (Bucentaur) superfamily is classified by an uncharacteristic conserved sequence of ∼80 amino acids (aa) at the C-terminus, BCNT-C (the conserved C-terminal region of Bcnt/Cfdp1). Whereas the yeast Swc5 and Drosophila Yeti homologues play crucial roles in chromatin remodelling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service