Skip to Content
Merck
All Photos(1)

Key Documents

SCT141

Sigma-Aldrich

BioTracker 540 Red Lysosome Dye

Live cell imaging dye for acidic cellular organelles such as lysosomes.

Sign Into View Organizational & Contract Pricing

Select a Size

SCT141
€300.00

€300.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
SCT141
€300.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

€300.00


Estimated to ship on05 May 2025Details


form

liquid

technique(s)

cell based assay: suitable

detection method

fluorometric

General description

Lysosomes are membrane-enclosed organelles that contain an array of enzymes capable of breaking down all types of biological material including proteins, nucleic acids, carbohydrates, and lipids.
Lysosomes function as the digestive system of the cell, serving both to degrade material taken up from outside the cell and to digest obsolete components of the cell itself.
BioTracker Lysosome dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step.

The BioTracker 540 Red Lysosome Dye can be detected using the red Cy3 channel.

Spectral Properties:
  • Absorbance: 541nm
  • Emission: 634nm

Research Category:
Cell Imaging

Application

Live cell fluorescent imaging

Packaging

50μl

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service