Skip to Content
Merck
All Photos(1)

Key Documents

SCT108

Sigma-Aldrich

BioTracker 655 Red Cytoplasmic Membrane Dye

Live cell imaging lipophilic carbocyanine membrane dye suitable for long-term fluorescent cell labeling and cell tracking studies.

Synonym(s):

Live cell imaging probe, Live cell membrane imaging probe

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€300.00

€300.00


Estimated to ship on04 June 2025Details



Select a Size

Change View
1 ML
€300.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

€300.00


Estimated to ship on04 June 2025Details


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Lipophilic carbocyanine dyes are widely used for labeling neurons in tissues by retrograde labeling, and to label membranes in a wide variety of cell types. The dyes are weakly fluorescent in aqueous phase, but become highly fluorescent in lipid bilayers. Staining is highly stable with low toxicity and very little dye transfer in between cells, making the dyes suitable for long-term cell labeling and tracking studies. When live cells are stained, the dyes label plasma membranes and also are taken up into endocytic compartments. Cells can be fixed either before or after staining, although permeabilization affects the staining pattern.

Unlike PKH dyes, BioTracker Cytoplasmic Membrane Dyes do not require a complicated hypoosmotic labeling protocol. They are ready-to-use dye delivery solutions that can be added directly to normal culture media to label suspended or adherent cells in culture.

Spectral Properties
Absorbance: 644nm
Emission: 665nm

Application

Live cell fluorescent imaging
Live cell imaging lipophilic carbocyanine membrane dye suitable for long-term fluorescent cell labeling and cell tracking studies.
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Quality

Spectral Properties
Absorbance: 644nm
Emission: 665nm

Physical form

Liquid

Storage and Stability

Store BioTracker 655 Red Cytoplasmic Membrane Dye at 2-8ºC. Protect From Light.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can this product be used to bind to cell membrane of human smooth muscle cells?

    1 answer
    1. This product is a lipophilic carbocyanine membrane dye, it is not species-specific. This compound is not tested specifically with human smooth muscle, however, there is no reason to suspect that this material would not be suitable.
      Please see the links below to review additional publications that may be helpful:
      Histochemistry 1992 May;97(4):329-33. doi: 10.1007/BF00270034. PMID: 1618647 https://pubmed.ncbi.nlm.nih.gov/1618647/
      ACS Omega 2020, 5, 20, 11829–11835 https://doi.org/10.1021/acsomega.0c01416 https://pubs.acs.org/doi/10.1021/acsomega.0c01416

      Please see the link below to review additional product information on the BioTracker Live Cell Stains and Dyes:
      https://www.sigmaaldrich.com/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/imaging-analysis-and-live-cell-imaging/organelle-dyes-and-stains

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service