Skip to Content
Merck
All Photos(1)

Key Documents

SCM083

Sigma-Aldrich

ES2N Basal Medium

The ES2N Basal Medium is a defined serum-free neural basal medium.

Sign Into View Organizational & Contract Pricing

Select a Size

245 ML
€144.00

€144.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
245 ML
€144.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

€144.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Quality Level

form

liquid

technique(s)

cell culture | stem cell: suitable

input

sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type neural stem cell(s)

shipped in

dry ice

General description

ES2N Basal Medium is a defined serum-free neural basal medium. This base medium offers flexibility to customers adding their own neural supplements or growth factors. Must be supplemented prior to use.

Application

• When supplemented with Millipore’s serum-free Neuro2 and Neuro27 supplements this medium supports the monolayer differentiation of mouse ES and iPS cells to neurons.
• When supplemented with bFGF, EGF and the serum-free Neuro2 and Neuro27 supplements this medium supports the maintenance and expansion of neural stem cells.

Components

ES2N Basal Medium: (Cat. No. SCM083) One (1) 245 mL bottle containing standard base medium.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service