Skip to Content
Merck
All Photos(2)

Key Documents

S4030

Millipore

Dextran Sulfate Solution

50% (w/v) liquid

Synonym(s):

dextran sulfate buffer, hybridization accelerant, hybridization additive, northern blot hybridization component, southern blot hybridization component

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€275.00

€275.00


Estimated to ship on29 April 2025Details



Select a Size

Change View
100 ML
€275.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.52

€275.00


Estimated to ship on29 April 2025Details


Product Name

Dextran Sulfate 50% Solution,

packaging

pkg of 100 mL

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

hybridization: suitable (nucleic acid)

shipped in

ambient

General description

Dextran Sulfate (MW >500,000) is used as an additive in probe hybridization solutions. Its presence accelerates the rate of hybridization by 10 fold. The addition of Dextran Sulfate favors the formation of probe networks, resulting in greater sensitivity during membrane-bound nucleic acid analysis. The product is supplied as a 50% (w/v) solution.

Application

Dextran Sulfate - 50% Solution

To prepare a solution containing 10% Dextran Sulfate, add 8 ml of hybridization ingredients to 2 ml of 50% Dextran Sulfate Solution.



See the "Hybrisol Nomograph for Dextran Sulfate" link located under the "Protocols" tab at catalog number S4030 product specific page on www.chemicon.com for determining the proper concentration of dextran sulfate needed for various hybridization condition guidelines.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this in the form of dextran sulfate sodium salt?

    1 answer
    1. This product is prepared from Dextran Sulfate Sodium Salt, CAS 9011-18-1. The solution is not tested for sodium content during or following preparation.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service