Skip to Content
Merck
All Photos(1)

Key Documents

265152-M

Millipore

Dextran Sulfate, Sodium Salt, Molecular Biology Grade

Used to accelerate hybridization of DNA probes to immobilized nucleic acids.

Synonym(s):

Dextran Sulfate, Sodium Salt, Molecular Biology Grade

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€315.00

€315.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
€315.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

€315.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

grade

Molecular Biology

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 5 mg/mL

foreign activity

DNases, proteases, RNases, none detected

storage temp.

2-8°C

General description

A long chain polymer of sulfated glucose, containing 17 - 20% sulfur. Has been used as an anticoagulant and has been shown to inhibit the binding of HIV-1 to CD4+ T lymphocytes. Used to accelerate hybridization of DNA probes to immobilized nucleic acids.
Long chain polymer of glucose containing 17-20% sulfur. Has been used as an anticoagulant, and has been shown to inhibit the binding of HIV-1 to CD4+ T-lymphocytes. Used to accelerate hybridization of DNA probes to immobilized nucleic acids.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cabrera, C., et al. 1999. AIDS Res. Hum. Retroviruses15, 1535.
Wahl, G.M., et al. 1979. Proc. Natl. Acad. Sci. USA 76, 3683.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service