Key Documents
RDHSTAND1
Milli-Q
Stand for Remote Dispenser
For Direct-Q® and Synergy® remote systems
Select a Size
1 EA
€663.00
€663.00
Estimated to ship on10 April 2025Details
Request more information
Select a Size
Change View
1 EA
€663.00
About This Item
UNSPSC Code:
41104200
NACRES:
SB.51
€663.00
Estimated to ship on10 April 2025Details
Request more information
packaging
pkg of 1 unit
compatibility
for use with Direct-Q® UV
for use with Direct-Q®
for use with Synergy® UV
for use with Synergy®
General description
The versatile remote dispenser, when supported by this stand, can be placed on a bench up to 2 m from the water purification unit.
Application
For Direct-Q® or Synergy® remote systems.Provides the support to place the Remote Dispenser on a lab bench.
Legal Information
DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany