PCC300C15C
Pellicon® Capsule with Ultracel® Membrane
C Screen, 1.5 m2 AseptiQuik® G connectors
Synonym(s):
Pellicon® Capsule with Ultracel® Membrane
Select a Size
+32 2 620 02 27or
This item is not able to be purchased as a guest user. For further information, please contact us at .Select a Size
About This Item
+32 2 620 02 27or
This item is not able to be purchased as a guest user. For further information, please contact us at .Recommended Products
material
EPDM rubber seal
PPO/PS blend (core)
PPO/PS blend housing
TPE seal
composite regenerated cellulose (CRC) membrane
epoxy adhesive
polyester screen
polypropylene screen
polyurethane adhesive
Quality Level
sterility
sterile; γ-irradiated
product line
Pellicon®
feature
holdup volume 456 mL (in feed channel)
holdup volume 741 mL (in permeate channel)
packaging
package of 1
parameter
max. inlet pressure at 75 psi (5.1 bar) and 4-30 °C
max. transmembrane pressure at 45 psi (3.1 bar) and 4-30 °C
4-6 L/min-m2 flow rate
technique(s)
ultrafiltration: suitable
diam.
10.3 cm (4.1 in.)
filtration area
1.5 m2
screen size
, Type C screen (coarse screen)
size
43.7 cm (17.2 in.) , Length
impurities
≤5 ppm Meets TOC (in retentate effluent, after a WFI flush of 20 L/m²)
matrix
Ultracel®
pore size
300 kDa
pH range
2-13
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Permeate: AseptiQuik® G Connector
Application
Features and Benefits
- Plug ′n play, holderless design - easy to install, safe to remove
- Gamma Sterilized and preseravtive-free - ready to process in minutes
- True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
- Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
- Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families
Packaging
Other Notes
- Comprehensive documentation to support qualification, risk assessment and process optimization needs.
- Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
- Convenient 24/7 access to up-to-date product information.
Legal Information
related product
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service