PXC300C50
Pellicon® XL Cassette with Ultracel® Membrane
filtration area 50 cm2, NMWCO 300 kDa
Synonym(s):
Pellicon XL Ultrafiltration Module Ultracel 300 kDa 0.005 m2
Select a Size
€424.00
Estimated to ship on05 May 2025Details
Select a Size
About This Item
€424.00
Estimated to ship on05 May 2025Details
Recommended Products
material
composite regenerated cellulose (CRC) membrane
polypropylene
polypropylene screen
Quality Level
feature
holdup volume 3.2 mL
manufacturer/tradename
Pellicon®
parameter
0.03–0.05 L/min recirculation rate
15 mL min. working volume
2.8 bar max. transmembrane pressure (40 psig)
5.5 bar max. inlet pressure (80 psi) at 50 °C
technique(s)
analytical sample preparation: suitable
protein purification: suitable
L
18.8 cm (7.4 in.)
W
3 cm (1.2 in.)
filtration area
50 cm2
membrane nominal area
50 cm2
gravimetric extractables
≤7.5 mg/device
matrix
Ultracel®
pore size
300 kDa NMWCO
pH
2 –12
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
Packaging
Other Notes
Analysis Note
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service