P3C010C01
Pellicon® 3 Cassette with Ultracel® Membrane
filtration area 0.11 m2, NMWCO 10 kDa
Synonym(s):
Pellicon® 3 Cassette with Ultracel 10 kDa Membrane, C screen, 0.11 m2
About This Item
Recommended Products
material
TPE seal
composite regenerated cellulose (CRC)
polypropylene cap (for end)
polypropylene screen
Quality Level
feature
holdup volume 18 mL (in feed channel)
manufacturer/tradename
Pellicon®
parameter
30 psi max. reverse press. exposure at 25 °C (for 24 hours)
4-6 L/min-m2 recirculation rate
40 psi max. transmembrane pressure at 4-50 °C
6.9 bar max. inlet pressure (100 psi)
80 psi max. transmembrane pressure at 4-40 °C
technique(s)
analytical sample preparation: suitable
buffer exchange: suitable
protein purification: suitable (concentration)
H
2.3 cm (0.90 in.)
L
20.6 cm (8.1 in.)
W
5.6 cm (2.2 in.)
filtration area
0.11 m2
membrane area
0.11 m2
gravimetric extractables
≤1500 mg/m2
matrix
Ultracel® (Membrane)
pore size
10 kDa NMWCO
pH
2-13
Looking for similar products? Visit Product Comparison Guide
General description
The Pellicon® 3 Cassette with Ultracel® Membrane is designed for the filtration of therapeutic proteins (such as monoclonal antibodies) and recombinant and non-recombinant proteins.
This P3C010C01 Pellicon® 3 ultrafiltration device features a pore size of 10 kDa NMWCO, a filtration area of 0.11 m2, a PLCGC membrane, and dimensions measuring 0.8 cm (H) x 5.6 cm (W) x 20.6 cm (L).
Features and Benefits
- Rugged & reliable – ideally suited for today′s more demanding filtration processes
- High performance – produces excellent resolution, yields and back-pressure resistance
- Optimum product recovery – thanks to proven membrane technology
- Void-free technology – leading-edge membranes resolve virtually any separation challenge
- Defect-free, superior adhesion – Ultracel®uses composite membranes cast on a microporous substrate
- Automated manufacturing – delivers unbeatable performance, consistency and reliability
- Easy installation & care – simple to install and maintain
Other Notes
Analysis Note
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service