Skip to Content
Merck

P3C005C10

Millipore

Pellicon® 3 Cassette with Ultracel® Membrane

5 kDa, C screen, 1.14 m2

Synonym(s):

Pellicon® 3 Cassette with ULTRACEL® 5 kDa Membrane, C Screen, 1.14 m2

Sign Into View Organizational & Contract Pricing

Select a Size

P3C005C10
€7,326.00

€7,326.00


Estimated to ship on14 April 2025Details



Select a Size

Change View
P3C005C10
€7,326.00

About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602

€7,326.00


Estimated to ship on14 April 2025Details


material

cap (Polypropylene-encapsulated stainless steel end cap)
composite regenerated cellulose (CRC) membrane
polypropylene (Screen (feed))
polypropylene housing

Quality Level

product line

Pellicon®

storage condition

do not freeze

parameter

4-8 L/min-m2 flow rate

technique(s)

tangential flow filtration (TFF): suitable
ultrafiltration: suitable

H

2.4 cm (0.93 in.)

L

20.6 cm (8.1 in.)

filtration area

1.14 m2

size

68 mL , Volume (permeate)
85 mL , Volume (feed)

width

17.8 cm (7 in.)
178 mm

matrix

(Ultracel)

pore size

5 kDa pore size

pH

2-13

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Pellicon® 3 cassettes with Ultracel® membrane are advanced, high-performance cassettes that are ideal for today’s higher titer therapeutic antibodies, as well as for the more demanding filtration processes that require higher operating pressures, temperatures, concentrations and caustic cleaning regimes.
From small-scale to full-scale production, Pellicon® 3 cassettes are designed for use in research, process scale-up/scale-down, applications development and full-scale manufacturing. The Pellicon® 3 cassette design and automated manufacturing process provides unbeatable performance consistency and enhanced linear scalability between cassette sizes. Pellicon® 3 cassettes also offer greater cassette size selection for improved scale-up and scale-down process development. The streamlined design allows operators to quickly and easily handle, install and remove Pellicon® 3 cassettes. The materials of construction are compatible with a broad range of chemical cleaning agents that many TFF systems require to ensure proper sanitization.

Application

monoclonal antibodies, recombinant and non-recombinant proteins, vaccines

Features and Benefits

  • Optimum product recovery using proven composite membrane technology
  • Fast, reliable scale-up/-down from lab to production scale
  • Rugged, reliable design ideally suited to filtration processes with higher operating pressures, temperatures and caustic cleaning regimes
  • Automated manufacturing delivers unbeatable performance consistency and reliability
  • Easy to install and clean
  • Extreme temperature and chemical compatibility
  • Choice of screens to best optimize your process

Other Notes

These products are manufactured in a facility which adheres to Good Manufacturing Practices.

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service