Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 each | Estimated to ship on29 September 2026fromMerck Life Science BV/SRL | €4,590.00 |
About This Item
€4,590.00
Estimated to ship on29 September 2026Details
Quality Segment
material
cap (Polypropylene-encapsulated stainless steel end cap), composite regenerated cellulose (CRC) membrane, polypropylene (Screen (feed)), polypropylene housing
product line
Pellicon®
storage condition
do not freeze
parameter
4-8 L/min-m2 flow rate
technique(s)
tangential flow filtration (TFF): suitable, ultrafiltration: suitable
H
2.7 cm (1.07 in.)
L
20.6 cm (8.1 in.)
filtration area
0.57 m2
size
127 mL , Volume (feed), 39 mL , Volume (permeate)
width
17.8 cm (7 in.) , 178 mm
matrix
(Ultracel)
pore size
30 kDa pore size
pH
1-14
storage temp.
2-8°C
General description
filtration processes that require higher operating pressures, temperatures and
caustic cleaning regimes.
From small-scale to full-scale production, Pellicon® 3 cassettes are designed for use in research, process scale-up/scale-down, applications development and full-scale manufacturing. The Pellicon® 3 design and automated manufacturing process provides unbeatable performance consistency between cassette sizes. Pellicon® 3 devices also offer greater cassette size selection for improved scale-up and scale-down process development. The streamlined design allows operators to quickly and easily handle, install and remove Pellicon® 3 cassettes. The materials of construction are compatible with a broad range of chemical cleaning.
Application
Features and Benefits
- Robust, void-free membrane provides optimum product retention and performance consistency
- Unique feed screen design enables high mass transfer, flux for higher final concentration
- Fast, reliable scale up/down from lab to production scale
- Automated manufacturing delivers unbeatable performance consistency and reliability
- Easy to install and clean
- Extreme temperature and chemical compatibility
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description 30 kDa, D screen, 0.57 m2 | description 30 kDa, D screen, 0.11 m2 | description 30 kDa, D screen, 88 cm2 | description 5 kDa, C screen, 1.14 m2 |
| pore size 30 kDa pore size | pore size 30 kDa pore size | pore size 30 kDa pore size | pore size 5 kDa pore size |
| product line Pellicon® | product line Pellicon® | product line Pellicon® | product line Pellicon® |
| technique(s) tangential flow filtration (TFF): suitable, ultrafiltration: suitable | technique(s) ultrafiltration: suitable, tangential flow filtration (TFF): suitable | technique(s) ultrafiltration: suitable, tangential flow filtration (TFF): suitable | technique(s) tangential flow filtration (TFF): suitable, ultrafiltration: suitable |
| filtration area 0.57 m2 | filtration area 0.11 m2 | filtration area 88 cm2 | filtration area 1.14 m2 |
| matrix (Ultracel) | matrix (Ultracel) | matrix (Ultracel) | matrix (Ultracel) |
| material polypropylene (Screen (feed)), cap (Polypropylene-encapsulated stainless steel end cap), composite regenerated cellulose (CRC) membrane, polypropylene housing | material polypropylene cap (end cap), polypropylene (Screen (feed)), polypropylene housing, composite regenerated cellulose (CRC) membrane | material composite regenerated cellulose (CRC) membrane, polypropylene (Screen (feed)), polypropylene housing, polypropylene cap (end cap) | material cap (Polypropylene-encapsulated stainless steel end cap), composite regenerated cellulose (CRC) membrane, polypropylene housing, polypropylene (Screen (feed)) |
Still not finding the right product?
Explore all of our products under Pellicon® 3 Cassette with Biomax® Membrane
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
wgk
WGK 2
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
64-19-7
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Difficulty Finding Your Product Or Lot/Batch Number?
How to Find the Product Number
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.

