Skip to Content
Merck
All Photos(1)

Key Documents

OP43L

Sigma-Aldrich

Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1)

lyophilized, clone DO-1, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€585.00

€585.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€585.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€585.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DO-1, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

feline, human

should not react with

mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG2a

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with X63Ag8.653 mouse myeloma cells (see application references). Recognizes the ~53 kDa mutant and wild-type forms of p53.
Recognizes the ~53 kDa wild-type and mutant p53 protein in A431 cells and breast carcinoma tissue.
This Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1) is validated for use in Frozen sections, Gel Shift, Immunoblotting, ICC, IP, Paraffin sections for the detection of p53 (Ab-6) (Pantropic).

Immunogen

Epitope: Within amino acids 21-25 of human p53
Human
wild type, recombinant, human p53

Application

Frozen sections (1 µg/ml; see application references)

Gel Shift (see comments)

Immunoblotting (0.1 µg/ml; see application references)

Immunocytochemistry (1-2.5 µg/ml; see application references)

Immunoprecipitation (1 µg/ml or use Cat. No. OP43A; see application references)

Paraffin sections (1 µg/ml, pepsin, heat or pressure cooker pre-treatment required; see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 100 µg BSA.

Reconstitution

We recommend resuspending the lyophilized antibody with sterile phosphate buffered saline (PBS), pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 μg/ml. Reconstituted product will be more stable if 0.1% sodium azide is added (do not add azide if antibody is to be used with viable cells). Lyophilized antibodies should be resuspended at 4°C with occasional gentle mixing for at least two hours. Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage.

Analysis Note

Negative Control
SK-OV-3 cells or normal skin tissue
Positive Control
A431 cells or breast carcinoma tissue

Other Notes

El-Deiry, W.S., et al. 1994. Cancer Res.54, 1169.
Greenblatt, M.S., et al. 1994. Cancer Res.54, 4855.
Legros, Y., et al. 1994. Oncogene9, 2071.
Barak, Y., et al. 1993. EMBO J.12, 461.
Kastan, M.B., et al. 1992. Cell71, 587.
Kuerbitz, S.J. 1992. Proc. Natl. Acad. Sci. USA89, 7491.
Lane, D.P. 1992. Nature358, 15.
Vojtesek, B., et al. 1992. J. Immunol. Meth.151, 237.
Kastan, M.B., et al. 1991 Cancer Res.51, 6304.
Recognizes both mutant and wild-type p53 under denaturing and non-denaturing conditions. This antibody reacts weakly with rodent p53; we do not recommend it for rodent samples. For gel shift assay, resuspend in 100 µl of buffer. Wild-type p53 has a short half life and is present in low amounts in cells. For immunoprecipitation increasing the amount of sample and labeling with 35S-Met for less than or equal to 1 h will aid in visualizing wild-type p53. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darren M Hutt et al.
Methods in molecular biology (Clifton, N.J.), 1510, 77-91 (2016-10-21)
Gene expression is regulated in part through the reversible acetylation of histones, by the action of histone acetyltransferases (HAT) and histone deacetylases (HDAC). HAT activity results in the addition of acetyl groups on the lysine residues of histone tails leading
E E Hoskins et al.
Oncogene, 27(35), 4798-4808 (2008-04-29)
Fanconi anemia (FA) is a genome instability syndrome that is characterized by progressive bone marrow failure and a high risk of cancer. FA patients are particularly susceptible to leukemia as well as squamous cell carcinomas (SCCs) of the head and
Masaki Kobayashi et al.
Oncotarget, 11(18), 1653-1665 (2020-05-15)
Poly (ADP-ribose) polymerase 1 (PARP1) plays important roles in single strand DNA repair. PARP1 inhibitors enhance the effects of DNA damaging drugs in homologous recombination-deficient tumors including tumors with breast cancer susceptibility gene (BRCA1) mutation. Nutlin-3a, an analog of cis-imidazoline
Sindy Sohr et al.
Cell cycle (Georgetown, Tex.), 10(21), 3758-3767 (2011-10-29)
Successful pregnancy requires a functionally normal blastocyst encountering a receptive maternal endometrium. Interestingly, the cell cycle regulator and tumor suppressor p53 has been reported to support reproduction in mice by regulating the expression of the leukemia inhibitory factor gene in
Yan Degenhardt et al.
Molecular cancer therapeutics, 9(7), 2079-2089 (2010-06-24)
Polo-like kinases are a family of serine threonine kinases that are critical regulators of cell cycle progression and DNA damage response. Predictive biomarkers for the Plk1-selective inhibitor GSK461364A were identified by comparing the genomics and genetics of a panel of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service