Skip to Content
Merck
All Photos(4)

Key Documents

MXRP2SPRKT

Millipore

Milliflex® Rapid 2.0 AutoSpray Station kit, with no supply

Rapid detection,response and resolution of microbial contamination

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€26,040.00

€26,040.00


Available to ship on17 April 2025Details


Request more information

Select a Size

Change View
1 EACH
€26,040.00

About This Item

UNSPSC Code:
41116129
NACRES:
JA.13

€26,040.00


Available to ship on17 April 2025Details


Request more information

AC/DC input

110 V / 230 V

material

316L stainless steel holder (for membrane)
316L stainless steel holder (for pad)
316L stainless steel turntable(s)
polyester key
polyurethane case
polyurethane cover

packaging

pkg of 1 kit

manufacturer/tradename

Milliflex®

parameter

15-40 °C temperature

technique(s)

Rapid Sterility Testing: suitable

H

31 cm (12 in.)

depth

40 cm (101.6 in.)

weight

10 kg (22 lb)

input

sample type pharmaceutical(s)

application(s)

bioburden testing
food and beverages
pharmaceutical
sterility testing
water monitoring

compatibility

for use with Milliflex® Rapid

detection method

bioluminescent

shipped in

ambient

General description

An integral component in the Milliflex® Rapid System 2.0, the Milliflex® Rapid 2.0 AutoSpray Station applies an equal volume of two reagents across a membrane surface in order to detect living microorganisms. Using a proven, pre-set automated program, the AutoSpray Station ensures reproducible results, test after test. Reagent application is achieved using a back and forth sweeping movement of the filter holder under the calibrated glass nebulizer. In automatic spray mode, the AutoSpray Station delivers both reagents in 90 seconds at the touch of a button.
From the control panel, analysts can easily initiate an operation by selecting from a list of operations.
Pinch valves ensure that the same volume of reagent is dispensed during each cycle. An internal compressor allows stand-alone operation, eliminating the need for additional equipment.

Application

Rapid detection,response and resolution of microbial contamination

Components

  • Includes: AutoSpray station
  • Filter holder
  • Pad holder
  • Nebulizer cleaner
  • 5ml female syringe
  • 3 nebulizers
  • silicone liquid tubes
  • 2 stainless steel caps

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Use Milliflex® Rapid System to detect cell-based immunotherapeutics for microbial contaminants. This automated ATP-based system supports rapid detection, imaging, and quantification of viable microorganisms.

Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.

Related Content

Milliflex® Rapid system delivers bioburden and sterility testing results days earlier with proven technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service