Skip to Content
Merck
All Photos(1)

Key Documents

MXRPBLRST

Millipore

Milliflex® Rapid Reagent Kit

Pharmaceutical Products Analysis

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€667.00

€667.00


Estimated to ship on27 June 2025Details


Request more information

Select a Size

Change View
100 TESTS
€667.00

About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NA.85

€667.00


Estimated to ship on27 June 2025Details


Request more information

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

input

sample type pharmaceutical(s)

application(s)

food and beverages
pharmaceutical

compatibility

Milliflex® Rapid

shipped in

wet ice

General description

An integral component in the Milliflex Rapid Microbiology Detection and Enumeration system, the Milliflex Rapid AutoSpray Station applies an equal volume of two reagents across a membrane surface in order to detect living microorganisms.
Using a proven, pre-set automated program, the AutoSpray Station ensures reproducible results, test after test. Reagent application is achieved using a back and forth sweeping movement of the filter holder under the calibrated glass nebulizer.

Using a proven, pre-set automated program, the AutoSpray Station ensures reproducible results, test after test. Reagent application is achieved using a back and forth sweeping movement of the filter holder under the calibrated glass nebulizer.
Using a proven, pre-set automated program, the AutoSpray Station ensures reproducible results, test after test. Reagent application is achieved using a back and forth sweeping movement of the filter holder under the calibrated glass nebulizer.

Using a proven, pre-set automated program, the AutoSpray Station ensures reproducible results, test after test. Reagent application is achieved using a back and forth sweeping movement of the filter holder under the calibrated glass nebulizer.

Pinch valves ensure that the same volume of reagent is dispensed during each cycle. An internal compressor allows stand-alone operation, eliminating the need for additional equipment.
In automatic spray mode, the AutoSpray Station delivers both reagents in 90 seconds at the touch of a button.
From the control panel, analysts can easily initiate an operation by selecting from a list of operations

Components

Includes: ATP releasing agent, 2bottles

Bioluminescent reagent, 2bottles

Reconstitution buffer, sterile, 2bottles

2 syringes

5Luer-Lok caps

6vial adapters

Analysis Note

Target Microorganisms: Total aerobic microorganisms

Pseudomonas species

Yeast and Mold

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Use Milliflex® Rapid System to detect cell-based immunotherapeutics for microbial contaminants. This automated ATP-based system supports rapid detection, imaging, and quantification of viable microorganisms.

Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.

Related Content

Milliflex® Rapid system delivers bioburden and sterility testing results days earlier with proven technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service