Skip to Content
Merck
All Photos(1)

Key Documents

MABT338

Sigma-Aldrich

Anti-phospho-cytokeratin-8 (K8) (Ser431) Antibody, clone 5B3

clone 5B3, from mouse

Synonym(s):

Keratin, type II cytoskeletal 8, CK-8, Keratin-8, K8, Type-II keratin Kb80

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€353.00

€130.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5B3, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pSer431)

Gene Information

human ... KRT8(3856)
mouse ... Krt8(16691)

General description

Keratin, type II cytoskeletal 8 (UniProt: P05787; also known as Cytokeratin-8, CK-8 Keratin-8, K8, Type-II keratin Kb80) is encoded by the KRT8 (also known as CYK8) gene (Gene ID: 3856) in human. Cytokeratin-8 is a type II, neutral to basic, protein of the intermediate filament family that together with Cytokeratin-19 (KRT19) helps to link the contractile apparatus to dystrophin at the costameres of striated muscle. Cytokeratin-8 can undergo phosphorylation on three major serine residues: Serine 23, 431, and 73. Serine 23 is shown to be highly conserved in all type II keratins. Phosphorylation at Serine 431 is reported to increase during mitosis and upon treatment with epidermal growth factor. Cytokeratin-8 can also undergo O-glycosylation in a cell cycle-dependent manner and glycosylation increases its solubility and reduces stability by inducing proteasomal degradation. Mutations in KRT8 gene have been linked to liver cirrhosis that is characterized by severe panlobular liver-cell swelling with Mallory body formation, prominent pericellular fibrosis, and marked deposits of copper. Two isoforms of Cytokeratin-8 have been described that are produced by alternative splicing.

Specificity

Clone 5B3 specifically detects cytokeratin-8 phosphorylated on serine 431.

Immunogen

Cytokeratin-8/18 purified from okadaic acid-treated HT29 cells.

Application

Anti-phospho-cytokeratin-8 (K8) (Ser431), clone 5B3, Cat. No. MABT338, is a mouse monoclonal antibody that detects type II Cytokeratin-8 phopshorylated on serine 431 and has been tested for use in Immunofluorescence and Western Blotting.
Immunofluorescence Analysis: A representative lot detected phospho-cytokeratin-8 (K8) (Ser431) in Immunofluorescence applications (Ku, N.O., et. al. (1997). J Biol Chem. 272(11):7556-64).

Western Blotting Analysis: A representative lot detected phospho-cytokeratin-8 (K8) (Ser431) in Western Blotting applications (Yoon, K.H., et. al. (2001). J Cell Biol. 153(3):503-16; Ridge, K.M., et. al. (2005). J Biol Chem. 280(34):30400-5; Ku, N.O., et. al. (1997). J Biol Chem. 272(11):7556-64).

Quality

Evaluated by Western Blotting in lysates from HeLa cells treated with Calyculin A/Okadaic Acid.

Western Blotting Analysis: 2 µg/mL of this antibody detected phospho-cytokeratin-8 (K8) (Ser431) in lysate from HeLa cells treated with Calyculin A/Okadaic Acid.

Target description

~53 kDa observed; 53.70 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service