Skip to Content
Merck
All Photos(1)

Key Documents

L1043

Sigma-Aldrich

10x Single Cell Lysis & Fragmentation Buffer

Sign Into View Organizational & Contract Pricing

Select a Size

500 REACTIONS
€66.40

€66.40


Please contact Customer Service for Availability


Select a Size

Change View
500 REACTIONS
€66.40

About This Item

UNSPSC Code:
12161703
NACRES:
NA.25

€66.40


Please contact Customer Service for Availability

Quality Level

shipped in

wet ice

storage temp.

−20°C

Application

Recommended for use with the SeqPlex DNA Amplification Kits

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saori Takahashi et al.
Nature genetics, 51(3), 529-540 (2019-02-26)
Here, we report a single-cell DNA replication sequencing method, scRepli-seq, a genome-wide methodology that measures copy number differences between replicated and unreplicated DNA. Using scRepli-seq, we demonstrate that replication-domain organization is conserved among individual mouse embryonic stem cells (mESCs). Differentiated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service