Skip to Content
Merck
All Photos(2)

Key Documents

MABT106

Sigma-Aldrich

Anti-Protein Spindly Antibody, clone 9D10.2

clone 9D10.2, from mouse

Synonym(s):

Protein Spindly, hSpindly, Arsenite-related gene 1 protein, Coiled-coil domain-containing protein 99, Rhabdomyosarcoma antigen MU-RMS-40.4A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9D10.2, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SPDL1(54908)

General description

Protein spindly, otherwise known as the coiled-coil domain-containing protein 99 (CCDC99), is a phosphoprotein that regulates mitosis. It interacts with the Rod/ZW10/Zwilch complex, and is involved in recruiting, docking, and alignment of motor protein dynein at mitotic kinetochores. Spindly may also be involved in checkpoint signaling and microtubule attachment.

Immunogen

GST-tagged recombinant protein corresponding to human Protein Spindly.

Application

Detect Protein Spindly using this Anti-Protein Spindly Antibody, clone 9D10.2 validated for use in ICC, Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Protein Spindly in HeLa and NIH/3T3.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected Protein Spindly in 10 µg of HEK293 cell lysate.

Target description

~70 kDa observed. This protein is known to exist in two isoforms at 70 kDa and 56 kDa An uncharacterized band may be observed at ~38 kDa in some cell lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingteng Xie et al.
Cell research, 28(4), 462-475 (2018-02-22)
Before fertilization, mammalian oocyte undergoes an asymmetric division which depends on eccentric positioning of the spindle at the oocyte cortex to form a polar body and an egg. Since the centriole is absent and, as a result, the polar array

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service