Skip to Content
Merck
All Photos(2)

Key Documents

MABS1994M

Sigma-Aldrich

Anti-MT-ND4 Antibody, clone 9E4-2D8

clone 9E4-2D8, from mouse

Synonym(s):

NADH-ubiquinone oxidoreductase chain 4, EC: 1.6.5.3, NADH dehydrogenase subunit 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€317.00

€317.00


Estimated to ship on12 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on12 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E4-2D8, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MT-ND4(4538)

General description

NADH-ubiquinone oxidoreductase chain 4 (UniProt: P03905; also known as EC: 1.6.5.3, NADH dehydrogenase subunit 4, MT-ND4) is encoded by the MT-ND4 (also known as MTND4, NADH4, ND4) gene (Gene ID: 4538) in human. MT-ND4 is an inner mitochondrial membrane protein that belongs to the complex I subunit 4 family. It is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). The complex couples the oxidation of NADH and the reduction of ubiquinone (Coenzyme Q10), to the generation of a proton gradient, which is then used for ATP synthesis. The complex can be dissociated into two main sub-complexes, corresponding to the "ankle" of the boot, and the "foot" of the boot. The ankle is thought to protrude from the membrane so as to be predominantly in the aqueous phase on the matrix side. It contains the binding site for NAD(H), and the input electron transfer chain. The foot (hydrophobic) is membrane bound and contains a catalytic site at which ubiquinone is reduced. Defects in MT-ND4 gene are known to cause Leber hereditary optic neuropathy (LHON) that is characterized by acute or subacute loss of central vision due to optic nerve dysfunction. LHON results from primary mitochondrial DNA mutations affecting the respiratory chain complexes. Other diseases associated with MT-ND4 gene mutations are age-related macular degeneration, mesial temporal lobe epilepsy, and cystic fibrosis.

Specificity

Clone 9E4-2D8 detects NADH-ubiquinone oxidoreductase chain 4 in mitochodria isolated from human cells.

Immunogen

KLH-conjugated linear peptide from the C-terminal region.

Application

Anti-MT-ND4, clone 9E4-2D8, Cat. No. MABS1994,is a mouse monoclonal antibody that detects NADH-ubiquinone oxidoreductase chain 4 and has been tested for use in Western Blotting.
Western Blotting Analysis: 4 µg/mL from a representative lot detected MT-ND4 in mitochondria from Human Neonatal Dermal Fibroblasts and mitochondria from Human neonatal dermal fibroblasts depleted of mtDNA. (Courtesy of Michael F. Marusich, Ph.D., mAbDx, Inc., Eugene, OR, USA).

Quality

Evaluated by Western Blotting in Mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA.

Western Blotting Analysis: 1 µg/mL of this antibody detected MT-ND4 in Mitochondria from human neonatal dermal fibroblasts and did not detect it in mitochondria from human neonatal dermal fibroblasts depleted of mtDNA.

Target description

~37 kDa observed; 51.58 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG2a in buffer containing HEPES-Buffered Saline (150 mM NaCl, 15 mM HEPES, pH 7.2) with 0.02% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Sheng Zhang et al.
Nature cell biology, 23(7), 684-691 (2021-07-14)
Members of the mammalian AlkB family are known to mediate nucleic acid demethylation1,2. ALKBH7, a mammalian AlkB homologue, localizes in mitochondria and affects metabolism3, but its function and mechanism of action are unknown. Here we report an approach to site-specifically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service