Skip to Content
Merck
All Photos(2)

Key Documents

MABS1132

Sigma-Aldrich

Anti-CDKL5 Antibody, clone 8F3.1

clone 8F3.1, from mouse

Synonym(s):

Cyclin-dependent kinase-like 5, Serine/threonine-protein kinase 9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8F3.1, monoclonal

species reactivity

mouse, rat

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cdkl5(382253)

General description

Cyclin-dependent kinase-like 5 (EC 2.7.11.22; UniProt O76039; also known as Serine/threonine-protein kinase 9) is encoded by the CDKL5 (also known as EIEE2, STK9) gene (Gene ID 6792) in human. CDKL5/STK9 is a serine/threonine protein kinase widely distributed in all tissues, with highest levels in thymus, testes, and brain, where it is found localized in the nucleus and dendrite of neurons. CDKL5/STK9 expression is found peaked in the early postnatal period and Cdkl5-knockdown in mice leads to autistic-like behavior, dendritic hypotrophy and impairment of neural circuit communication. Likewise, human CDKL5 gene mutations are linked to X-linked neurological developmental disorders involving severe mental retardation and seizures appearing in the first postnatal months. Point mutations in CDKL5 have also been reported in patients with atypical variants of Rett syndrome (RTT), infantile spasms and severe neurodevelopmental retardation.

Specificity

Expected to react with both spliced isoforms of human CDKL5 reported by UniProt (O76039).

Immunogen

Epitope: Internal.
GST-tagged recombinant human CDKL5 internal fragment.

Application

Anti-CDKL5 Antibody, clone 8F3.1 is an antibody against CDKL5 for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected CDKL5 in 10 µg of rat hippocampus tissue lysate.

Quality

Evaluated by Western Blotting in mouse cerebellum tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CDKL5 in 10 µg of mouse cerebellum tissue lysate.

Target description

~115 kDa observed. 115.5 kDa (isoform 1) and 107.5 kDa (isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Young Kim et al.
Nature communications, 11(1), 1924-1924 (2020-04-23)
Renal tubular epithelial cells (RTECs) perform the essential function of maintaining the constancy of body fluid composition and volume. Toxic, inflammatory, or hypoxic-insults to RTECs can cause systemic fluid imbalance, electrolyte abnormalities and metabolic waste accumulation- manifesting as acute kidney

Questions

  1. Which region was the mAb raised against for the CDKL6 protein?

    1 answer
    1. The immunogen for MABS1132 is a GST-tagged recombinant human CDKL5 internal fragment. It is located within the 400-550 amino acid range, and the precise epitope has not been characterized.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service