Skip to Content
Merck
All Photos(2)

Key Documents

MABS1140

Sigma-Aldrich

Anti-Fgr Antibody, clone SuGr-7 14F3

clone SuGr-7 14F3, from mouse

Synonym(s):

Tyrosine-protein kinase Fgr, Fgr, Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog, Proto-oncogene c-Fgr, p55-Fgr, p58-Fgr, p58c-Fgr

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€296.00

€296.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€296.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€296.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SuGr-7 14F3, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FGR(2268)

General description

Tyrosine-protein kinase Fgr (EC 2.7.10.2; UniProt P09769; also known as c-fgr, c-src2, c-src-2 proto-oncogene, Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog, p55-Fgr, p58-Fgr, p58c-Fgr, proto-oncogene c-Fgr) is encoded by the FGR (also known as SRC2) gene (Gene ID 2268) in human. Fgr is a member of the Src family of non-receptor protein tyrosine kinases (PTKs). Fgr contains N-terminal myristylation and palmitylation sites, a PTK catalytic domain, as well as SH2 and SH3 domains mediating interactions with phosphotyrosine-containing and proline-rich motifs, respectively. Fgr is a positive regulator of mast cell activation/degranulation and is essential for IgE-mediated passive cutaneous anaphylaxis (PCA) in mice. Fgr appears to act cooperatively with Fyn in the activation of Syk and its downstream targets in a manner that is counteracted by Lyn.

Immunogen

Recombinant protein corresponding to human Fgr.

Application

Detect FGR using this mouse monoclonal Anti-Fgr Antibody, clone SuGr-7 14F3, Cat. No. MABS1140, validated for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Fgr in 10 µg of Ramos cell lysate.

Quality

Evaluated by Western Blotting in Daudi cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Fgr in 10 µg of Daudi cell lysate.

Target description

~59 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service