Skip to Content
Merck
All Photos(2)

Key Documents

MABT1333

Sigma-Aldrich

Anti-pan-Actin Antibody

mouse monoclonal, 2A3

Synonym(s):

Skeletal muscle alpha-actin, Smooth muscle alpha-actin, Cytoplasmic beta-actin, Cardiac muscle alpha-actin, Cytoplasmic gamma-actin, Smooth muscle gamma-actin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
100 μL
€417.00

€153.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
100 μL
€417.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€153.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Product Name

Anti-pan-Actin Antibody, clone 2A3, clone 2A3, from mouse

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A3, monoclonal

species reactivity

human, rat, monkey, yeast, hamster, Drosophila, mouse

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Actin is a multi-functional cytoskeletal protein that serves as the basic building block of cytoskeletal microfilaments. Six different actin types have been reported. Skeletal muscle alpha-actin (UniProt: P68133) is encoded by the ACTA1 gene, smooth muscle alpha-actin (UniProt: P62736) by the ACTA2 gene, cytoplasmic beta-actin (UniProt: P60709) by the ACTB gene, cardiac muscle alpha-actin (UniProt: P68032) by the ACTC1 gene, cytoplasmic gamma-actin (UniProt: P63261) by the ACTG1 gene, and smooth muscle gamma-actin (UniProt: P63267) by the ACTG2 (also known as ACTA3) gene. Although actins show about 90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expression patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic beta and gamma actins are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt. This pan-actin antibody detects all isoforms actin.

Specificity

Clone 2A3 detects all forms of actin in multiple species. It targets an epitope with in the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to a fragment from the C-terminal region of human Actin.

Application

Anti-pan-Actin, clone 2A3, Cat. No. MABT1333, is a mouse monoclonal antibody that detects all forms of actin and has been tested for use in Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: A 1:50 dilution from a representative lot detected Actins in various species samples (e.g. Human, rat, mouse, monkey, Hamster, Drosophila, Yeast) (Courtesy of Martina Mitterhuber, M.sc and Egon Ogris, M.D., Medical University of Vienna, Austria).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Actins in HeLa cell lysate.

Target description

~42 kDa observed; 41.74 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Zhao et al.
Cancer immunology research, 6(12), 1459-1471 (2018-09-14)
Although anti-PD-1 therapy has improved clinical outcomes for select patients with advanced cancer, many patients exhibit either primary or adaptive resistance to checkpoint inhibitor immunotherapy. The role of the tumor stroma in the development of these mechanisms of resistance to
Congxin Sun et al.
Cell reports, 42(5), 112372-112372 (2023-04-22)
Autophagy is a homeostatic process critical for cellular survival, and its malfunction is implicated in human diseases including neurodegeneration. Loss of autophagy contributes to cytotoxicity and tissue degeneration, but the mechanistic understanding of this phenomenon remains elusive. Here, we generated
Hung-Hsi Chen et al.
iScience, 24(9), 103086-103086 (2021-09-28)
Using antibody arrays, we found that the RNA helicase DDX3 modulates the expression of secreted signaling factors in oral squamous cell carcinoma (OSCC) cells. Ribo-seq analysis confirmed amphiregulin (AREG) as a translational target of DDX3. AREG exerts important biological functions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service