Skip to Content
Merck
All Photos(1)

Key Documents

MABN2485

Sigma-Aldrich

Anti-Cdc42 Antibody, clone 28-10

clone 28-10, from mouse

Synonym(s):

Cell division control protein 42, Suppressor of RHO3 protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00
100 μL
€353.00

€130.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00
100 μL
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

28-10, monoclonal

species reactivity

yeast, human, mouse

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CDC42(998)

General description

Cell division control protein 42 (UniProt: P19073; also known as Cdc42, Suppressor of RHO3 protein 2) is encoded by the CDC42 (also known as SRO2, TLR229C, L8083.13) gene (Gene ID: 850930) in yeast (Saccharomyces cerevisiae). Cdc42 is a member of the Rho GTPase family that regulates multiple signaling pathways involved in actomyosin organization, cell proliferation, and maintenance of cell polarity in most eukaryotic cell types. Cdc42 signaling process involves guanine nucleotide exchange factors (GEFs), guanine nucleotide dissociation inhibitors (GDIs), and GTPase-activating proteins (GAPs). In yeast, generation and maintenance of robust Cdc42 polarity promotes membrane expansion during bud formation and it has been shown that Cdc42 is dynamically maintained at the polarity cap through its continuous cycling between the polarity cap and internal pools. Cdc42 is highly polarized during bud emergence and is specifically required for exocytosis at this stage. Cdc42 is synthesized with a propeptide region (aa 189-191), which is cleaved off in the mature form. The mature form contains three nucleotide (GTP)-binding sites. (Ref.: Wu, H., and Brennwald, P. (2010) Mol. Cell Biol. 30(21); 5207-5217).

Specificity

Clone 28-10 specifically detects Cell division control protein 42 (Cdc42).

Immunogen

GST-tagged full-length recombinant yeast Cdc42.

Application

Anti-Cdc42, clone 28-10, Cat. No. MABN2485, is a highly specific mouse monoclonal antibody that targets Cell division control protein 42 (Cdc42) and has been tested for use in Immunocytochemistry and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected Cdc42 in Western Blotting applications (Sullivan, C.S., et. al. (2016). J Biol Chem. 291(51):26262-26272; Wu, H., et. al. (2010). Mol Cell Biol. 30(21):5207-17).

Immunocytochemistry Analysis: A representative lot detected Cdc42 in Immunocytochemistry applications (Wu, H., et. al. (2010). Mol Cell Biol. 30(21):5207-17).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Cdc42 in HeLa cell lysate.

Target description

~21 kDa observed; 21.32 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service