Skip to Content
Merck
All Photos(1)

Documents

MABF841

Sigma-Aldrich

Anti-FCGR2A/C Antibody, clone 11B6

clone 11B6, from mouse

Synonym(s):

Low affinity immunoglobulin gamma Fc region receptor II-a, Low affinity immunoglobulin gamma Fc region receptor II-b, Low affinity immunoglobulin gamma Fc region receptor II-c, IgG Fc receptor II-a, IgG Fc receptor II-b, IgG Fc receptor II-c, CDw32, Fc-g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11B6, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

FcγRI/CD64, FcγRII/CD32, FcγRIII/CD16, and FcγRIV/CD16-2 represent the four known classes of IgG Fc receptors (FcγRs). FcγRII, FcγRIII, and FcγRIV are low-affinity receptors for monomeric IgG, whereas FcγRI is the only high-affinity FcγR. FcγRs play important roles in inflammatory cell activation, clearance, presentation of Ag, and maintenance of IgG homeostasis. In addition to binding immune complex (IC), FcγRs have been shown to bind non-IgG ligands. For example, FcγRII is shown to bind oxLDL, while FcγRIII binding to an E. coli component is reported to negatively regulate the function of macrophage receptor with collagenous structure (MARCO). FcyRII activation is also reported to transduce a negative regulatory signal against CD38 cross-linking-induced proliferation of resting mature B cells. Low affinity immunoglobulin gamma Fc region receptor II-a/b/c (IgG Fc receptor II-a/-b/-c, CDw32, Fc-gamma RII-a/-b/-c, Fc-gamma-RIIa/b/c, FcRII-a/-b/-c, CD32) are encoded by the FCGR2A/B/C (CD32, FCG2, IGFR2) genes in human (UniProt P12318, P31994, P31995; Gene ID 2212, 2213, 9103). The FcγRIIs encoded by the three genes are highly conserved in their extracellular portion, with significant variation in the transmembrane and intracellular regions. All three types of transcripts alternative splicing to generate additional isoforms.

Specificity

Clone 11B6 recognizes human FcγRIIa and FcγRIIc, but not FcγRIIb, by targeting a conserved cytoplasmic domain sequence in human FcγRIIa and FcγRIIc.

Immunogen

Epitope: Cytoplasmic domain.
Linear peptide corresponding to a cytoplasmic domain sequence identical in human FcγRIIa and FcγRIIc.

Application

Anti-FCGR2A/C Antibody, clone 11B6 is an antibody against FCGR2A/C for use in Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blotting.
Immunocytochemistry Analysis: A representative lot detected FcγRIIc immunoreactivity in paraformaldehyde-fixed, Triton X-100-permeabilized human B cell lines carrying the FCGR2C-ORF allele, but not B cell lines with homozygous FCGR2C-STP alleles (Li, X., et al. (2013). Sci. Transl. Med. 5(216):216ra175).
Western Blotting Analysis: A representative lot detected recombinant FcγRIIc cytoplasmic (CY) domain GST fusion, but not FcγRIIb CY domain GST fusion (Li, X., et al. (2013). Sci. Transl. Med. 5(216):216ra175).
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Evaluated by Immunohistochemistry in human bone marrow tissue.

Immunohistochemistry Analysis: An 1:50 dilution of this antibody detected FCGR2A/C in human bone marrow tissue.

Target description

30.89 kDa calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan J Hubbard et al.
The Journal of experimental medicine, 217(10) (2020-07-14)
IgG immune complexes (ICs) promote autoimmunity through binding fragment crystallizable (Fc) γ-receptors (FcγRs). Of these, the highly prevalent FcγRIIa (CD32a) histidine (H)-131 variant (CD32aH) is strongly linked to human autoimmune diseases through unclear mechanisms. We show that, relative to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service