Skip to Content
Merck
All Photos(1)

Key Documents

MABF3228

Sigma-Aldrich

Anti-Complement C3/C3b Antibody, clone 10C7

Synonym(s):

PZP-like alpha-2-macroglobulin domain-containing protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€137.00
100 μL
€347.00

€137.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€137.00
100 μL
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€137.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10C7, monoclonal

mol wt

calculated mol wt 187.15 kDa

purified by

using protein G

species reactivity

mouse, human

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
flow cytometry: suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... C3(718)

Specificity

Clone 10C7 is a mouse monoclonal antibody that detects Complement C3b.

Immunogen

Purified complement C3 from human serum.

Application

Quality Control Testing

Evaluated by ELISA with purified native Human Complement C3

ELISA Analysis: Serial dilutions of this antibody detected Human Complement C3, but not Complement C4 protein.

Tested Applications

ELISA Analysis: A representative lot detected Complement C3/C3b in ELISA applications (Whipple, E.C., et al. (2007). Mol Immunol. 44(4):377-88).

Flow Cytometry Analysis: A representative lot detected Complement C3/C3b in Flow Cytometry applications (Sharma, S., et al. (2020). Front Immunol.11:462).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Complement C3 (UniProt: P01024; also known as C3, PZP-like alpha-2-macroglobulin domain-containing protein 1) is encoded by the C3 (also known as CPAMD1) gene (Gene ID: 718) in human. C3 is a secreted protein that plays a key role in the activation of the complement system. It s processing by C3 convertase is the central reaction in both classical and alternative complement pathways. C3 has an anaphylatoxin-like domain (aa 693-728) and a NTR (Netrin) domain (aa 1518-1661). IC3 precursor is first processed by the removal of four arginine residues, forming two chains, β and , linked by a disulfide bond. C3 convertase between residues 748 and 749 activates C3 by cleaving the chain, releasing C3a anaphylatoxin and generating C3b. C3a appears to be important in many inflammatory responses. The active C3b fragment covalently binds to the cell or bacterial surface carbohydrates or immune aggregates, via its reactive thioester, and plays a role in opsonization. Surface-associated C3b generated in the classical pathway can also feed into the alternative pathway because it has the affinity for binding with the Factor B to form a pro-convertase C3bB. C3a anaphylatoxin is a mediator of local inflammatory process. In chronic inflammation, it acts as a chemoattractant for neutrophils. Defects in C3 gene can cause complement component 3 deficiency that leads to recurrent, severe, pyogenic infections because of ineffective opsonization of pathogens. Defects in C3 gene are also known to cause age-related macular degeneration and hemolytic uremic syndrome that can lead to hemolytic anemia and renal failure Clone 10C7 specifically detects human and mouse C3b. (Ref.: Sharma, S., et al. (2020). Front. Immunol. 11; 462; Whipple, EC., et al. (2007). Mol. Immunol 44(4); 377-388).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service