Skip to Content
Merck
All Photos(1)

Documents

MABF2733

Sigma-Aldrich

Anti-Chikungunya virus E2 Antibody, clone CHK-152

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

CHK-152, monoclonal

mol wt

calculated mol wt 138.07 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
inhibition assay: suitable
neutralization: suitable

isotype

IgG2κ

epitope sequence

Unknown

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

General description

Chikungunya virus (CHIKV) that causes distinctive polyarthritis or polyarthralgia with clinical features such as fever, maculopapular rash, and myalgia is transmitted by Aedes spp of mosquitoes. Immune compromised individuals may face serious complications, including encephalitis and mortality. CHIKV genome contains a single-stranded positive-sense RNA that encodes four non-structural proteins known as nsP1, nsP2, nsP3, and nsP4 and also five structural proteins that include a capsid protein, three envelope glycoproteins known as (E1, E2, and E3, and a small molecule known as 6K. The mature alphavirus particles express E1 and E2 heterodimers that form 80 trimeric spikes on the surface of the virion. The mature E2 protein contains three domains with immunoglobulin-like folds: N-terminal domain A, domain B at the tip; and the C-terminal domain C located proximal to the viral membrane. The ectodomain E1 protein consists of three domains known as D1, DII, and DIII. The DIII domain is an immunoglobulin-like domain connected to D1 and DII by a flexible linker. Both E1 and E2 proteins are responsible for virus entry into host cells. The E2 glycoprotein interacts with a cellular receptor, resulting in the virus internalization and the E1 glycoprotein mediates virus fusion to host cell under low pH conditions. Following the fusion of the viral envelope with the endosomal membrane, the viral genomic RNA is released into the cytoplasm and starts replicating. Clone CHK-152 is a humanized antibody that blocks viral fusion and is shown to protect mice against CHIKV-induced mortality and disease. (Ref.: Zhang, R., et al. (2019). Cell Reports 28(10); 2647-2658; Pal, P., et al. (2013). PLoS Pathog. 9(4); e1003312).

Specificity

Clone CHK-152 is a humanized mouse monoclonal antibody that specifically detects E2 protein in Chikungunya virus. It targets an epitope on A domain of E2.

Immunogen

Irf-7 deficient mice injected with La Reunion 2006 OPY-1 strain of Chikungunya virus (CHIKV-LR) and boosted with CHIKV virus-like particles and E2 protein.

Application

Quality Control Testing

Evaluated by ELISA with recombinant Chikungunya Virus E2 Envelope Protein.

ELISA Analysis: Various dilutions of this antibody detected recombinant Chikungunya virus E2 protein.

Tested Applications

Surface plasmon resonance: A representative lot detected Chikungunya virus E2 in Surface plasmon resonance applications (Pal, P., et al. (2013). PLoS Pathog. 9(4):e1003312).

Neutralizing Analysis: A representative lot neutralized CHIKV and protected immunocompromised mice against disease. (Pal, P., et al. (2013). PLoS Pathog. 9(4):e1003312).

ELISA Analysis: A representative lot detected Chikungunya virus E2 in ELISA applications (Zhang, R., et al. (2019). Cell Rep. 28(10):2647-2658.e5).

Inhibition Analysis: A representative lot inhibited CHIKV infection when added after virus adsorption to cell surface. (Pal, P., et al. (2013). PLoS Pathog. 9(4):e1003312).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Chikungunya virus E2, clone CHK-152, Cat. No. MABF2733, is a mouse monoclonal antibody that detects E2 protein in Chikungunya virus (CHIKV) and is tested for use in ELISA, Inhibition, Neutralizing, and Surface plasmon resonance.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service