The molecular weight for the antibody is approximately 150 kDa, according to our team.
SAB5700589
Monoclonal Anti-2019-nCoV Spike antibody produced in Rabbit
Select a Size
€389.00
Select a Size
About This Item
€389.00
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody product type
primary antibodies
clone
monoclonal
form
liquid
species reactivity
SARS virus
technique(s)
ELISA: 1:1000-1:5000
flow cytometry: 1:50-1:200
immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:200
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
SARS virus ... S(43740568)
General description
Storage and Stability
Disclaimer
Data presented is the available current product information and provided as-is. These products have not been tested or verified in any additional applications, sample types, including any clinical use. Experimental conditions must be empirically derived by the user. Our Antibody Guarantee only covers tested applications stated herein and conditions presented in our product information and is not extended to publications.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the molecular weight (MW) of product SAB5700589?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service