Skip to Content
Merck
All Photos(2)

Key Documents

MABE164

Sigma-Aldrich

Anti-ID4 Antibody, clone DML07

clone DML07, from mouse

Synonym(s):

DNA-binding protein inhibitor ID-4, Class B basic helix-loop-helix protein 27, bHLHb27, Inhibitor of DNA binding 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DML07, monoclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ID4(3400)

General description

ID4 is an inhibitor of differentiation protein, which, along with ID1, ID2 and ID3, plays a key role in the regulation of lineage commitment, cell fate decisions and in the timing of differentiation during neurogenesis, lymphopoiesis and neovascularisation (angiogenesis). ID proteins, induced by the bone morphogenic protein-SMAD pathway, have also been shown to cooperate with LIF to ensure that embryonic stem cells opt for self-renewal.

Immunogen

KLH-conjugated linear peptide corresponding to human ID4.

Application

Use Anti-ID4 Antibody, clone DML07 (Mouse Monoclonal Antibody) validated in WB to detect ID4 also known as DNA-binding protein inhibitor ID-4, Class B basic helix-loop-helix protein 27, bHLHb27.
Western Blot Analysis: 1 µg/mL from a representative lot detected ID4 on 10 µg of mouse lymph node tissue lysate.

Quality

Evaluated by Western Blot in human lung tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected ID4 on 10 µg of human lung tissue lysate.

Target description

~17 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service