Skip to Content
Merck

1.08219

SAFC

Tris(hydroxymethyl)aminomethane hydrochloride

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€337.00
2.25 KG
€485.00
5 KG
€1,301.00
10 KG
€2,322.00
12 KG
€2,781.00
50 KG
€10,345.00

€337.00


Estimated to ship on07 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€337.00
2.25 KG
€485.00
5 KG
€1,301.00
10 KG
€2,322.00
12 KG
€2,781.00
50 KG
€10,345.00

About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
MDL number:
UNSPSC Code:
12352107
EC Index Number:
214-684-5

€337.00


Estimated to ship on07 April 2025Details


Request a Bulk OrderRequest a Sample

vapor pressure

<1 Pa ( 50 °C)

Quality Level

product line

EMPROVE® EXPERT

form

solid

feature

RNase and DNase free

pH

4.2 (20 °C, 100 g/L in H2O)

useful pH range

7.0-9.0

pKa (25 °C)

8.1

mp

150.7 °C

solubility

soluble 561 g/L

density

1.28 g/cm3 at 20.2 °C

application(s)

parenterals

storage temp.

15-25°C

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right chemical that matches your pharma and biopharma manufacturing needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for pharma and biopharma manufacturing withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Other Notes

Due to its low microbial and endotoxin limits, Tris(hydroxymethyl)aminomethane hydrochloride Emprove® Expert is suitable for biopharma manufacturing needs.
The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service