Skip to Content
Merck
All Photos(3)

Key Documents

MABC522

Sigma-Aldrich

Anti-MAP17 Antibody, clone 165C

clone 165C, from mouse

Synonym(s):

PDZK1-interacting protein 1, 17 kDa membrane-associated protein, Protein DD96

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

165C, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

The protein named PDZK1-interacting protein 1 or alternatively 17kDa membrane-associated protein DD96 or MAP17 and encoded by the human gene named PDZK1IP1 or MAP17 is a highly unusual protein being naturally expressed only in a single epithelial cell population found in the proximal tubular epithelial cells. However, MAP17 is expressed in cancerous tissue of the kidney, colon, lung and breast. MAP17 expression in cancers causes an increase in reactive oxygen species production and tumorigenesis in cancerous tissues. These factors are mediated though MAP17’s PDZ-binding domain because disruption of this sequence by point mutations abolishes the ability of MAP17 to enhance ROS production and tumorigenesis. MAP17 also often serves as a cancer cell marker to distinguish tumor margins and tumorigenesis potential. MAP17 is a membrane bound protein and is thought to be a putative oncogene but its exact function in normal cells is still being researched.

Immunogen

GST-tagged recombinant protein corresponding to human MAP17.

Application

Anti-MAP17 Antibody, clone 165C is a highly specific mouse monoclonal antibody, that targets MAP17 & has been tested in western blotting & IHC.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected MAP17 in 10 µg of human salivary gland tissue lysate.
Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected MAP17 in human prostate cancer and human kidney tissue.

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MAP17 in 10 µg of human kidney tissue lysate.

Target description

~20 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service