Skip to Content
Merck
All Photos(1)

Key Documents

1.09034

Supelco

Schiff′s reagent

for detection of aldehydes

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€74.60

€74.60


Estimated to ship on02 July 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
1 L
€74.60

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21
grade:
for analytical purposes
form:
liquid

€74.60


Estimated to ship on02 July 2025Details


Request a Bulk OrderRequest more information

grade

for analytical purposes

Quality Level

form

liquid

technique(s)

photometry: suitable

pH

1.5 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

storage temp.

15-25°C

General description

For the analytical determination of aldehydes in the ppm range. For histology we recommend Schiff′s reagent item 109033

Application


  • Macrophagic myofasciitis lesions assess long-term persistence of vaccine-derived aluminium hydroxide in muscle.: This study examines the long-term persistence of vaccine-derived aluminum hydroxide in muscle, identified using special staining techniques including Schiff′s reagent, to evaluate the chronic effects on macrophagic myofasciitis lesions. (Gherardi RK et al., 2001).

Analysis Note

Assay (SO₂): 0.15 - 0.30 %
Application test: conforms

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service