Skip to Content
Merck
All Photos(1)

Key Documents

MABC1824

Sigma-Aldrich

Anti-phospho-MYO10 (Ser1060/1062/1066) Antibody, clone 2C10-6

Synonym(s):

Unconventional myosin-10, Unconventional myosin-X

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€137.00
100 μL
€347.00

€137.00


Estimated to ship on06 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€137.00
100 μL
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€137.00


Estimated to ship on06 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C10-6, monoclonal

mol wt

calculated mol wt 237.35 kDa
observed mol wt ~260 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... MYO10(4651)

General description

Unconventional myosin-X (UniProt: Q9HD67; also known as Unconventional myosin-10, MYO10) is encoded by the MYO10 (also known as KIAA0799) gene (Gene ID:4651) in human. Myosins are highly conserved, ubiquitously expressed proteins that form regular bipolar thick filaments that together with actin thin filaments constitute the fundamental contractile unit of skeletal and cardiac muscle. Along with actin they serve as motor molecules with ATPase activity that is essential for muscle contraction. MYO10, a multi-domain protein, is ubiquitously expressed and is present as a monomer in the cytosol in an inactive conformation. However, it is homodimeric in its active, membrane-bound conformation. It interacts strongly with calmodulin 3 (CALM3) and this interaction is essential for its function in filopodial extension and motility. It contains a MyTH4 and an FERM domain at its C-terminal, which are required for its binding to tubulin and β-integrin, respectively. The N-terminal region contains a sequence (aa 619-641) that is required for its bind to actin. Its myosin motor domain is localized in amino acids 63-739. MYO10 is reported to promote tumor growth through inducing genomic instability and chronic inflammation in cancer cells. It regulates genomic instability, at least partially, through mediating mitotic progression. MYO10 is reported to contain a Degron motif, a short stretch of amino acids that drives the degradation of unstable protein. In addition, phosphorylation of serine residues in MYO10 (serine 1060, 1062, 1066) drives its β-TrCP1-dependent degradation. Phosphorylated form of MYO10 is shown to be less stable. Degron mutation are reported to abolish MYO10 phosphorylation, degradation, and mitotic regulation. Cancer cells expressing high levels or stabilized MYO10 display higher sensitivity to Taxol. Deletion of Degron motif results in complete stability of MYO10. (Ref.: Pozo, MF., et al. (2023). Cell Rep. 42(5); 112531; Pozo, MF., et al. (2021). Sci Adv. 7(38); eabg6908).

Specificity

Clone 2C10-6 is a mouse monoclonal antibody that detects MY10 phosphorylated on serine 1060, 1062, and 1066.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphoserines 1060, 1062, and 1066 from human MYO10.

Application

Quality Control Applications

Evaluated by Western Blotting in lysate U20S cells transfected with GFP-MYO10.

Western Blotting Analysis (WB): A 1:250 dilution of this antibody detected phospho-MYO10 (Ser 1060/1062/1066) in U20S cells stably transfected with GFP-MYO10, but not in lysate from cells with MYO10 knockout.

Tested applications

Immunocytochemistry Analysis: A representative lot detected p-MYO10 (Ser1060/1062/1066) in Immunocytochemistry application (Pozo, F. M., et al. (2021). Sci Adv. 7(38); eabg6908).

Western Blotting Analysis: A representative lot detected p-MYO10 (Ser1060/1062/1066) in Western Blotting application (Pozo, F. M., et al. (2021). Sci Adv. 7(38); eabg6908).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service