Skip to Content
Merck
All Photos(1)

Key Documents

MAB8671F

Sigma-Aldrich

Anti-Influenza B Antibody, Core, clone 22D5-12-13, FITC-conjugated

clone 22D5-12-13, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€542.00

€542.00


Estimated to ship on13 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€542.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

€542.00


Estimated to ship on13 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

22D5-12-13, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Influenza B, anti-core specific; reacts with all strains of Influenza B tested. No reactivity with Flu A or other respiratory viruses

Application

Anti-Influenza B Antibody, Core, clone 22D5-12-13, FITC-conjugated is an antibody against Influenza B for use in IF.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service