Skip to Content
Merck
All Photos(1)

Key Documents

MAB8671

Sigma-Aldrich

Anti-Influenza B Antibody, Core, clone 22D5-12-13

clone 22D5-12-13, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€555.00

€555.00


Estimated to ship on17 February 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€555.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€555.00


Estimated to ship on17 February 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

22D5-12-13, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Recognizes Influenza B, anti-core specific; reacts with all strains of Influenza B tested. No reactivity with Flu A or other respiratory viruses.

Immunogen

Epitope: Core

Application

Indirect Immunofluorescence

Optimal dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Influenza B Antibody, Core, clone 22D5-12-13 is validated for use in IF for the detection of Influenza B.

Physical form

Format: Purified
Protein A Purified. Liquid in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6 with 0.1% sodium azide
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Influenza Control Slides, Catalogue Number 5010-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of different cell substrates on the measurement of human influenza virus neutralizing antibodies.
Zhai, W; Zhang, DN; Mai, C; Choy, J; Jian, G; Sra, K; Galinski, MS
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service