Skip to Content
Merck
All Photos(1)

Key Documents

MAB8257F-5

Sigma-Aldrich

Anti-Influenza A Antibody, nucleoprotein, clone A1

clone A1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€1,550.00

€1,550.00


Estimated to ship on28 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZMS1020

Request a Bulk Order

Select a Size

Change View
500 μG
€1,550.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

€1,550.00


Estimated to ship on28 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZMS1020

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

A1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Specific for the Influenza A nucleoprotein. Has stronger binding with N1-type Flu A. No cross reactivity seen to influenza B or other respiratory viruses.

Immunogen

Epitope: nucleoprotein
Influenza A

Application

Indirect Immunofluorescence

Optimal dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Influenza A Antibody, nucleoprotein, clone A1 is validated for use in IF for the detection of Influenza A.

Physical form

Purified, FITC conjugated Liquid in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6 with 15 mg/mL BSA and 0.1% sodium azide

Storage and Stability

Store at 2° to 8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madeleine F Jennewein et al.
iScience, 25(4), 104088-104088 (2022-04-12)
Pregnancy represents a unique tolerogenic immune state which may alter susceptibility to infection and vaccine response. Here, we characterized humoral immunity to seasonal influenza vaccine strains in pregnant and non-pregnant women. Although serological responses to influenza remained largely intact during
Jie Zhou et al.
EMBO molecular medicine, 11(1) (2018-12-01)
Glycine decarboxylase (GLDC) was prioritized as a candidate susceptibility gene to severe influenza in humans. The higher expression of GLDC derived from genetic variations may confer a higher risk to H7N9 and severe H1N1 infection. We sought to characterize GLDC
Jeannine A Majde et al.
Journal of neurovirology, 13(5), 399-409 (2007-11-13)
Influenza pneumonitis causes severe systemic symptoms in mice, including hypothermia and excess sleep. The association of extrapulmonary virus, particularly virus in the brain, with the onset of such disease symptoms has not been investigated. Mature C57BL/6 male mice were infected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service