Skip to Content
Merck
All Photos(1)

Key Documents

MAB5346

Sigma-Aldrich

Anti-RAR alpha Antibody, clone 76

clone 763, Chemicon®, from mouse

Synonym(s):

RARalpha

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€446.00

€446.00


Estimated to ship on19 March 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1369

Request a Bulk Order

Select a Size

Change View
50 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on19 March 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1369

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

763, monoclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RARA(5914)

Specificity

Reacts with Retinoic Acid Receptor, alpha isotype. By Western blot MAB5346 recognizes a ~48 kDa band in rat brain hippocampal homogenate.

Immunogen

Peptide from the N-terminus of human Retinoic Acid Receptor, alpha isotype.

Application

Anti-RAR alpha Antibody, clone 763 is a Mouse Monoclonal Antibody for detection of Retinoic Acid Receptor also known as RAR alpha & has been validated in WB & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western blotting

Immunohistochemistry

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epidermal growth factor receptor (EGFR) is overexpressed in high-grade dysplasia and adenocarcinoma of the esophagus and may represent a biomarker of histological progression in Barrett's esophagus (BE).
Cronin, J; McAdam, E; Danikas, A; Tselepis, C; Griffiths, P; Baxter, J; Thomas, L; Manson et al.
The American Journal of Gastroenterology null
Joseph Mandelbaum et al.
The Journal of experimental medicine, 215(10), 2673-2685 (2018-09-14)
Pluripotent cells have been used to probe developmental pathways that are involved in genetic diseases and oncogenic events. To find new therapies that would target MYB-driven tumors, we developed a pluripotent zebrafish blastomere culture system. We performed a chemical genetic
Fred G Pluthero et al.
Methods in molecular biology (Clifton, N.J.), 1812, 13-31 (2018-09-02)
Microscopy is central to studies of platelets and their precursor megakaryocytes. Here we describe methods to rapidly obtain high resolution images of fixed platelets, megakaryocytes and megakaryocytic cells via immunofluorescence microscopy. Protocols covered include: (1) isolation and preparation of cells
Kohei Nishimoto et al.
Molecular metabolism, 40, 101025-101025 (2020-05-31)
Extrahepatic vitamin A is housed within organ-specific stellate cells that support local tissue function. These cells have been reported in the vocal fold mucosa (VFM) of the larynx; however, it is unknown how vitamin A reaches and is disseminated among

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service