Skip to Content
Merck
All Photos(2)

Key Documents

MAB2610

Sigma-Aldrich

Anti-Zyxin Antibody, clone 14E11.1

clone 14E11.1, from mouse

Synonym(s):

Zyxin2, Zyxin, Zyxin-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€379.00

€379.00


Estimated to ship on28 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1408

Request a Bulk Order

Select a Size

Change View
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship on28 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1408

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14E11.1, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Zyx(22793)

General description

Zyxin, is a cytosolic, focal adhesion protein belonging to the Ajuba family and containing 3 LIM zinc-binding domains. It associates with the actin cytoskeleton near the adhesion plaques and, during mitosis, with microtubules. Zyxin localizes to stress fibers, cell-cell junctions and colocalizes with vinculin, paxillin and VASP at focal adhesions but not at the tips of lamellipodia. Zyxin is involved in the regulation of actin dynamics and is a likely component of the signal transduction pathway mediating adhesion-stimulated changes in gene expression. Zyxin displays an actin nucleating activity that is dependent on its amino terminal proline-rich motifs and independent of Arp2/3 complex. Zyxin appears to function as an adaptor or docking protein to organize protein complexes important for regulated actin cytoskeleton dynamics.

Immunogen

KLH-conjugated linear peptide corresponding to human Zyxin.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a previous lot detected Zyxin in NIH/3T3, A431, and HeLa cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
This Anti-Zyxin Antibody, clone 14E11.1 is validated for use in WB, IC for the detection of Zyxin.

Quality

Evaluated by Western Blot in HUVEC cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Zyxin in 10 µg of HUVEC cell lysate.

Target description

~84 kDa observed. A non-specific band may be observed at ~36 kDa
The calculated molecular weight is 61 kDa Zyxin is a phosphoprotein with an apparent molecular weight of 82–84 kDa (van der Gaag, E., et al. (2002). Journal of Investigative Dermatology. 118:246–254.). As a substrate of caspases, Zyxin can be cleaved as 30, 37, 48 and 70 kDa (C-B Chan, C-B., et al. (2007). Cell Death and Differentiation. 14:1688–1699.)

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HUVEC cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Xu et al.
International journal of molecular sciences, 25(4) (2024-02-24)
Focal adhesions (FAs) play a crucial role in cell spreading and adhesion, and their autophagic degradation is an emerging area of interest. This study investigates the role of Thrombospondin Type 1 Domain-Containing Protein 1 (THSD1) in regulating autophagy and FA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service