Skip to Content
Merck
All Photos(3)

Key Documents

ABC1387M

Sigma-Aldrich

Anti-Zyxin

serum, from rabbit

Synonym(s):

Zyxin-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€317.00

€317.00


Estimated to ship on13 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on13 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ZYX(7791)

General description

Zyxin (UniProt: Q15942; also known as Zyxin-2) is encoded by the ZYX gene (Gene ID: 7791) in human. Zyxin is an evolutionarily conserved LIM domain protein that acts as an adhesion plaque protein concentrated at sites of cell adhesion and is widely expressed. It has been implicated in cell adhesion, cell motility, and signaling. Zyxin is reported to associate with members of the Enabled (Ena)/vasodilator-stimulated phosphoprotein (VASP) family of cytoskeletal regulators and plays a role in cytoskeletal dynamics and signaling. Zyxin contains three LIM zinc binding domains (aa 384-443; 444-503; and 504-570), which act as a negative regulator of Zyxin-VASP complexes. Also, it contains four proline rich ActA repeats (aa 64-77; 94-108; 115-121; and 127-137) that are known to interact directly with Ena/VASP proteins. Loss of Zyxin results in inability to thicken stress fibers in response to mechanical stimuli. (Ref.: Hoffman LM et al (2003). Mol. Cell Biol. 23(1); 70-79; Moody JD et al. (2009). Biochem. Biophys. Res. Commun. 378(3), 625-628; Yoshigi et al (2005). J. Cell Biol. 171(2), 209-215.).

Specificity

This rabbit polyclonal antibody detects Zyxin in human and murine cells. It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human Zyxin.

Application

Anti-Zyxin Antibody, Cat. No. ABC1387, is a rabbit polyclonal antibody that detects Zyxin in human and murine cells and has been tested for use in Immunocytochemistry and Western Blotting.
Western Blotting Analysis: A 1:5,000 dilution from a representative lot detected Zyxin in 10 µg of NIH/3T3 cell lysate.

Immunocytochemistry Analysis: A representative lot detected Zyxin in Immunocytochemistry applications (Hoffman, L.M., et. al. (2003). Mol Cell Biol. 23(1):70-9; Hoffman, L.M., et. al. (2006). J Cell Biol. 172(5):771-82).

Western Blotting Analysis: A representative lot detected Zyxin in Western Blotting applications(Hoffman, L.M., et. al. (2003). Mol Cell Biol. 23(1):70-9; Hoffman, L.M., et. al. (2006). J Cell Biol. 172(5):771-82).

Immunocytochemistry Analysis A 1:600 dilution from a representative lot detected Zyxin in mouse fibroblasts (Courtesy of Dr. Laura Hoffman at Huntsman Cancer Institute, University of Utah).

Western Blotting Analysis: A representative lot detected Zyxin in Western Blotting applications (Moody, J.D., et. al. (2009). Biochem Biophys Res Commun. 378(3):625-8).

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Zyxin in mouse fibroblast, HeLa, and U2OS cell lysates (Courtesy of Dr. Laura Hoffman at Huntsman Cancer Institute, University of Utah).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Zyxin in 10 µg of A431 cell lysate.

Target description

~75 kDa observed; 61.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service