Skip to Content
Merck
All Photos(3)

Documents

HNS2MAG-95K

Millipore

MILLIPLEX® Human Neuroscience Magnetic Bead Panel 2 - Neuroscience Multiplex Assay

The analytes available for this multiplex kit are: Angiogenin, ApoE4, FABP3, Ferritin, Neurogranin, and TREM2.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 124%
(Neurogranin)

accuracy: 84%
(FABP3)

accuracy: 85%
(ApoE4)

accuracy: 94%
(TREM2)

accuracy: 97%
(Angiogenin)

sensitivity: 11.0 pg/mL
(MinDC + 2SD; TREM2)

sensitivity: 14.3 pg/mL
(MinDC + 2SD; FABP3)

sensitivity: 162.8 pg/mL
(MinDC + 2SD; ApoE4)

sensitivity: 19.9 pg/mL
(MinDC + 2SD; Neurogranin)

sensitivity: 5.8 pg/mL
(MinDC + 2SD; Angiogenin)

sensitivity: 6.1 pg/mL
(MinDC + 2SD; Ferritin)

standard curve range: 15-50,000 pg/mL
(TREM2)

standard curve range: 2-10,000 pg/mL
(Angiogenin)

standard curve range: 24-100,000 pg/mL
(FABP3)

standard curve range: 244-1,000,000 pg/mL
(ApoE4)

standard curve range: 5-20,000 pg/mL
(Neurogranin)

standard curve range: 6-25,000 pg/mL
(Ferritin)

inter-assay cv: <10%
intra-assay cv: <5%
(Angiogenin)

inter-assay cv: <10%
intra-assay cv: <5%
(FABP3)

inter-assay cv: <10%
intra-assay cv: <5%
(TREM2)

inter-assay cv: <15%
intra-assay cv: <10%
(Ferritin)

inter-assay cv: <15%
intra-assay cv: <10%
(Neurogranin)

inter-assay cv: <15%
intra-assay cv: <5%
(ApoE4)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

General description

Monitoring protein biomarkers in cerebrospinal fluid (CSF) of patients with neurological disorders has been highly beneficial to understanding disease progression. While several CSF biomarkers can reproducibly distinguish normal and diseased samples, CSF is a difficult biological fluid to obtain in research studies. The need for blood-based biomarkers of AD has driven a continuous search for novel candidates. This configurable multiplex immunoassay quantitatively measures six proteins present in both human CSF and blood plasma/serum that are involved in neurological disease. Cell/tissue culture samples may also be used in the panel.

MILLIPLEX® Human Neuroscience Bead Panel 2 is a 6-plex kit to be used for the simultaneous quantification of any or all of the following analytes in cerebrospinal fluid (CSF), serum/plasma and cell/tissue culture supernatants for homogenates: Angiogenin, ApoE4, FABP3, Ferritin, Neurogranin, TREM2.

Panel Type: Neuroscience

Specificity

Cross Reactivty
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible. There is <5% cross-reactivity from ApoE2 and ApoE3 recombinant proteins on the ApoE4 beads.

Application

  • Analytes: Angiogenin, ApoE4, FABP3, Ferritin, Neurogranin, TREM2
  • Recommended Sample type: Plasma, Serum, CSF, Cell/Tissue culture supernatant
  • For Plasma/Serum a 1:10 dilution is recommended for the optimal detection of the analytes FABP3, Ferritin, Neurogranin and TREM2.
  • For Plasma/Serum a 1:200 dilution for ApoE4 and Angiogenin is recommended.
  • Because the sample dilutions differ for Plasma/Serum samples ApoE4 and Angiogenin cannot be plexed in the same assay as FABP3, Ferritin, Neurogranin and TREM2.
  • CSF samples require a 1:10 dilution and so may all be plexed together
  • Cell/Tissue culture supernatants or homogenates may be used neat or diluted in appropriate control medium.
  • Bead diluent must be present in order to detect Angiogenin. (Biotinylated-Angiogenin detection antibody is included in the bead diluent.)
  • Research Category: Neuroscience

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maciej Dulewicz et al.
Journal of clinical medicine, 10(19) (2021-10-14)
Synaptic loss and dysfunction are one of the earliest signs of neurodegeneration associated with cognitive decline in Alzheimer's disease (AD). It seems that by assessing proteins related to synapses, one may reflect their dysfunction and improve the understanding of neurobiological
Frederic Brosseron et al.
Journal of neurochemistry, 157(6), 2210-2224 (2020-09-08)
There is growing evidence that promising biomarkers of inflammation in Alzheimer´s disease (AD) and other neurodegenerative diseases correlate strongest to levels of tau or neurofilament, indicating an inflammatory response to neuronal damage or death. To test this hypothesis, we investigated

Related Content

MILLIPLEX® multiplex immunoassays detect Alzheimer's biomarkers in neuroscience research using high sensitivity technology.

Neuroscience multiplex assays measure biomarkers to understand neurodegenerative diseases, disorders, and neuropeptide signaling.

UnEnhance research with MILLIPLEX® multiplexing tips, industry guidance, and one-day assays for limited lab time.

Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service