Skip to Content
Merck
All Photos(2)

Key Documents

HATF09025

Millipore

Immobilon® MCE Membrane Filter

0.45 μm Pore Size, filter diam. 90 mm, hydrophilic

Synonym(s):

Immobilon®-NC Transfer Membrane

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€126.00

€126.00


Estimated to ship on10 June 2025Details



Select a Size

Change View
25 EA
€126.00

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

€126.00


Estimated to ship on10 June 2025Details


Product Name

MCE Membrane Filter, 0.45 μm Pore Size, Immobilon®, filter diam. 90 mm, hydrophilic

material

mixed cellulose esters (MCE) membrane (surfactant-free)
plain filter
white filter

Quality Level

description

90 mm diameter disc, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane

feature

hydrophilic

manufacturer/tradename

Immobilon®
Millipore

parameter

4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.

technique(s)

western blot: suitable

diam.

90 mm

filter diam.

90 mm

thickness

150 μm

color

white

refractive index

n/D 1.51

matrix

Immobilon®

pore size

0.45 μm pore size
79 % porosity

capacity

117 μg/cm2 (insulin)
160 μg/cm2 adsorption capacity (BSA)
259 μg/cm2 adsorption capacity (goat IgG)

bubble point

≥2.1 bar, air with water at 23 °C

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau-S red

detection method

colorimetric
fluorometric
radioactive

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

  • Biological solutions
  • Cell contact
  • Very small volumes requiring surfactant-free surfaces
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Wang et al.
Nephron, 144(2), 96-108 (2019-10-30)
Chloroquine (CQ), a classic autophagy inhibitor, is used clinically for malaria prophylaxis and pulmonary hypertension treatment. The adverse effects of CQ on morphological and functional changes in the kidney were investigated in the current study due to CQ accumulation in
Jiao Meng et al.
Annals of translational medicine, 8(5), 200-200 (2020-04-21)
Autophagy plays an important role in cellular homeostasis. Epigallocatechin gallate (EGCG), a polyphenol derived from green tea, has been shown to elicit vascular protective effects. Our study aimed to investigate the protective effect of EGCG in an endothelial injury model

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service