HATF01300
Immobilon® MCE Membrane Filter
0.45 μm Pore Size, filter diam. 13 mm, hydrophilic
Synonym(s):
Immobilon®-NC Transfer Membrane
Select a Size
€129.00
Estimated to ship on17 April 2025Details
Select a Size
About This Item
€129.00
Estimated to ship on17 April 2025Details
Recommended Products
Product Name
MCE Membrane Filter, 0.45 μm Pore Size, Immobilon®, filter diam. 13 mm, hydrophilic
material
mixed cellulose esters (MCE) membrane (surfactant-free)
plain filter
white filter
Quality Level
description
13 mm diameter disc, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane
feature
hydrophilic
manufacturer/tradename
Immobilon®
Millipore
parameter
4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.
technique(s)
western blot: suitable
diam.
13 mm
filter diam.
13 mm
thickness
150 μm
color
white
refractive index
n/D 1.51
matrix
Immobilon®
pore size
0.45 μm pore size
79 % porosity
capacity
117 μg/cm2 adsorption capacity (insulin)
160 μg/cm2 adsorption capacity (BSA)
259 μg/cm2 adsorption capacity (goat IgG)
bubble point
≥2.1 bar, air with water at 23 °C
compatibility
for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau-S red
detection method
colorimetric
fluorometric
radioactive
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Application
- Biological solutions
- Cell contact
- Very small volumes requiring surfactant-free surfaces
- Isolation of virus-like particles in wastewater
- Microplastics analysis grade water
- Nucleic acid binding, including eDNA
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service