Skip to Content
Merck
All Photos(5)

Key Documents

GFDX000800

Millipore

Glass Fiber Diagnostic Pad

filter W × L 8 mm × 100 m, pack of 1 roll

Synonym(s):

Conjugate pad, Diagnostic pad, Glass Fiber Diagnostic Pad 8 mm x 100 m roll, Glass fiber pad

Sign Into View Organizational & Contract Pricing

Select a Size

1 ROLL
€109.00

€109.00


Available to ship on15 April 2025Details


Request more information

Select a Size

Change View
1 ROLL
€109.00

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.23

€109.00


Available to ship on15 April 2025Details


Request more information

packaging

pack of 1 roll

Quality Level

manufacturer/tradename

SureWick®

L

100 m (328 ft)

W

8 mm

filter W × L

8 mm × 100 m

slit W

0.8 cm

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

SureWick® Pad Materials are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. SureWick® glass fiber conjugate / diagnostic pads are used to release detector reagents and faciltiate sample flow across the lateral flow membrane. This pack contains one 8mm x 100m roll of glass fiber diagnostic pad (GFDX) material.

Application

SureWick® Pad Materials are optimised for use as sample, conjugate, or absorbent pads in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.

Features and Benefits

  • SureWick® glass fiber pads have low extractables and excellent consistency.
  • They provide reliable, predictable sample flow and reagent delivery.
  • They exhibit consistent thickness, bed volume, and uniform surface quality.
  • Quick Delivery and Lead time.
  • Excellent Technical support

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service